Assay Details
Target Gene Details
Entrez Gene ID: | 67869 |
Gene Name: | polyadenylate-binding protein-interacting protein 2 |
Gene Aliases: |
2310050K10Rik, AU045972 |
Location: |
Chr.18:35598602-35617185 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paip2 | NM_026420.2 | NP_080696.1 | ||
| AK005462.1 | BAB24053.1 | |||
| AK009909.1 | BAB26578.1 | |||
| AK146327.1 | BAE27082.1 | |||
| AK150957.1 | BAE29989.1 | |||
| AK154453.1 | BAE32597.1 | |||
| AK167545.1 | BAE39611.1 | |||
| AK169207.1 | BAE40979.1 | |||
| AK169411.1 | BAE41157.1 | |||
| BC010190.1 | AAH10190.1 |

Genomic Map