Assay Details
Target Gene Details
Entrez Gene ID: | 67869 |
Gene Name: | polyadenylate-binding protein-interacting protein 2 |
Gene Aliases: |
2310050K10Rik, AU045972 |
Location: |
Chr.18:35598602-35617185 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paip2 | NM_026420.2 | 3 | 381 | NP_080696.1 |
| AK005462.1 | 3 | 383 | BAB24053.1 | |
| AK008252.1 | ||||
| AK009909.1 | 3 | 382 | BAB26578.1 | |
| AK146327.1 | 3 | 412 | BAE27082.1 | |
| AK150957.1 | 3 | 389 | BAE29989.1 | |
| AK154453.1 | 4 | 483 | BAE32597.1 | |
| AK167545.1 | 3 | 408 | BAE39611.1 | |
| AK169207.1 | 3 | 403 | BAE40979.1 | |
| AK169411.1 | 3 | 403 | BAE41157.1 | |
| BC010190.1 | 3 | 397 | AAH10190.1 |
Target Gene Details
Entrez Gene ID: | 20522 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 1 |
Gene Aliases: |
D18Ucla2, SVCT1, Slc23a2, YSPL3 |
Location: |
Chr.18:35604041-35630340 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc23a1 |

Genomic Map