Assay Details
Target Gene Details
Entrez Gene ID: | 17288 |
Gene Name: | meprin 1 beta |
Gene Aliases: |
Mep-1b |
Location: |
Chr.18:21072344-21100199 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mep1b | NM_008586.2 | NP_032612.2 | ||
| AK098127.1 | ||||
| BC125627.1 | ||||
| BC145979.1 | ||||
| L15193.1 | AAA75234.1 |
More Information
Set Membership: |
|

Genomic Map