Assay Details
Target Gene Details
Entrez Gene ID: | 621998 |
Gene Name: | predicted gene 6277 |
Gene Aliases: |
- |
Location: |
Chr.18:11821088-11839377 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6277 | NR_045421.1 | |||
| AK140316.1 |

Genomic Map