Assay Details
Target Gene Details
Entrez Gene ID: | 102466152 |
Gene Name: | microRNA 6357 |
Gene Aliases: |
Gm27333, mmu-mir-6357 |
Location: |
Chr.18:70516786-70516889 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6357 | NR_105775.1 |
Target Gene Details
Entrez Gene ID: | 26447 |
Gene Name: | polymerase (DNA directed), iota |
Gene Aliases: |
Rad30b |
Location: |
Chr.18:70508680-70530334 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poli | NM_001136090.2 | 9 | 1798 | NP_001129562.1 |
| NM_001289515.1 | 9 | 2443 | NP_001276444.1 | |
| NM_001289516.1 | 9 | 1729 | NP_001276445.1 | |
| NM_011972.2 | 9 | 1361 | NP_036102.2 | |
| AF151691.1 | 9 | 1361 | AAD50424.1 | |
| AK145244.1 | 9 | 1306 | BAE26322.1 | |
| AK154537.1 | 9 | 2392 | BAE32662.1 | |
| AY515316.1 | 9 | 1267 | AAS75834.1 | |
| AY515317.1 | 9 | 1267 | AAS75835.1 | |
| BC082278.1 | 9 | 1615 | AAH82278.1 | |
| HQ698885.1 | 9 | 1494 | ||
| HQ698886.1 | 9 | 1494 | ||
| HQ698887.1 | 9 | 1494 | ||
| HQ698888.1 | 9 | 1494 | ||
| HQ698889.1 | 9 | 1494 | ||
| HQ698890.1 | 9 | 1494 | ||
| HQ698891.1 | 9 | 1494 | ||
| HQ698892.1 | 9 | 1494 | ||
| HQ698893.1 | 9 | 1494 |
More Information
Set Membership: |
|

Genomic Map