Assay Details
Target Gene Details
Entrez Gene ID: | 69241 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide D |
Gene Aliases: |
2310002G05Rik, 2610028L19Rik, HSRBP4, RBP4 |
Location: |
Chr.18:31789159-31796701 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2d | NM_027002.3 | NP_081278.1 | ||
| NM_027101.2 | NP_081377.1 | |||
| AK009093.1 | ||||
| AK011600.1 | BAB27725.1 | |||
| BC004810.1 | AAH04810.1 |
More Information
Set Membership: |
|

Genomic Map