Assay Details
Target Gene Details
Entrez Gene ID: | 66388 |
Gene Name: | cutC copper transporter homolog (E.coli) |
Gene Aliases: |
2310039I18Rik, AI326282, CGI-32 |
Location: |
Chr.19:43753023-43768638 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cutc | NM_001113562.1 | 4 | 484 | NP_001107034.1 |
| NM_025530.3 | 4 | 484 | NP_079806.2 | |
| AK007591.1 | 4 | 409 | BAB25124.1 | |
| AK009702.1 | 4 | 509 | BAB26449.1 | |
| BC026775.1 | 4 | 394 | AAH26775.1 |
More Information
Set Membership: |
|

Genomic Map