Assay Details
Target Gene Details
Entrez Gene ID: | 56708 |
Gene Name: | cardiotrophin-like cytokine factor 1 |
Gene Aliases: |
Bsf3, CLC |
Location: |
Chr.19:4214392-4222615 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clcf1 | NM_019952.3 | NP_064336.1 | ||
| AF176913.1 | AAF00993.1 | |||
| AK170969.1 | BAE42148.1 |

Genomic Map