Assay Details
Target Gene Details
Entrez Gene ID: | 66583 |
Gene Name: | exosome component 1 |
Gene Aliases: |
2610035C18Rik, 2610104C07Rik, 2610312F07Rik, AI447561 |
Location: |
Chr.19:41922979-41933418 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exosc1 | NM_001164561.1 | NP_001158033.1 | ||
| NM_025644.4 | NP_079920.1 | |||
| AK002930.1 | BAB22465.1 | |||
| AK006018.1 | BAB24368.1 | |||
| AK011817.1 | ||||
| BC024423.1 | AAH24423.1 |
More Information
Set Membership: |
|

Genomic Map