Assay Details
Target Gene Details
Entrez Gene ID: | 104174 |
Gene Name: | glycine decarboxylase |
Gene Aliases: |
D030049L12Rik, D19Wsu57e |
Location: |
Chr.19:30098441-30175441 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gldc | NM_138595.2 | NP_613061.1 | ||
| AK080789.1 | BAC38022.1 | |||
| AK080937.1 | ||||
| AK139929.1 | ||||
| BC017135.1 | AAH17135.1 |
More Information
Set Membership: |
|

Genomic Map