Assay Details
Target Gene Details
Entrez Gene ID: | 14705 |
Gene Name: | Berardinelli-Seip congenital lipodystrophy 2 homolog (seipin) |
Gene Aliases: |
2900097C17Rik, AI046355, Gng3lg |
Location: |
Chr.19:8837467-8848683 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bscl2 | NM_001136064.3 | NP_001129536.1 | ||
| AK052341.1 | ||||
| BC061689.1 | AAH61689.1 | |||
| CK618551.1 |
Target Gene Details
Entrez Gene ID: | 14704 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 3 |
Gene Aliases: |
- |
Location: |
Chr.19:8836929-8839246 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng3 | NM_010316.3 | NP_034446.1 | ||
| AK004459.1 | BAB23313.1 | |||
| AK032646.1 | BAC27969.1 | |||
| BC029680.1 | AAH29680.1 | |||
| BY290040.1 | ||||
| U38497.1 | AAB01728.1 |
More Information
Set Membership: |
|

Genomic Map