Assay Details
Target Gene Details
Entrez Gene ID: | 269061 |
Gene Name: | cleavage and polyadenylation specific factor 7 |
Gene Aliases: |
5730453I16Rik, AL022757, C330017N18Rik |
Location: |
Chr.19:10525244-10547735 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf7 | NM_001164272.1 | NP_001157744.1 | ||
| NM_172302.3 | NP_758506.3 | |||
| AK053375.1 | BAC35368.1 | |||
| AK088603.1 | BAC40447.1 | |||
| AK165330.1 | BAE38138.1 | |||
| BC038812.1 | AAH38812.1 |
Target Gene Details
Entrez Gene ID: | 68642 |
Gene Name: | transmembrane protein 216 |
Gene Aliases: |
1110017C22Rik, 2810441K11Rik, 4921533J23Rik, A930021F15Rik, AI482550 |
Location: |
Chr.19:10539326-10556303 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem216 |
More Information
Set Membership: |
|

Genomic Map