Assay Details
Target Gene Details
Entrez Gene ID: | 69860 |
Gene Name: | eukaryotic translation initiation factor 1A domain containing |
Gene Aliases: |
2010003J03Rik, AI131644 |
Location: |
Chr.19:5366813-5371511 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1ad | NM_027236.2 | NP_081512.1 | ||
| AK008070.1 | BAB25440.1 | |||
| AK031807.1 | ||||
| AK083659.1 | 2 | 1468 | ||
| AK153194.1 | BAE31794.1 | |||
| AK158977.1 | BAE34752.1 | |||
| AK168102.1 | BAE40073.1 | |||
| AK168253.1 | BAE40203.1 | |||
| AK168598.1 | BAE40465.1 | |||
| BC024640.1 | AAH24640.1 |
More Information
Set Membership: |
|

Genomic Map