Assay Details
Target Gene Details
Entrez Gene ID: | 14227 |
Gene Name: | FK506 binding protein 2 |
Gene Aliases: |
13kDa, FKBP-2, mFKBP13, mFKBP2 |
Location: |
Chr.19:6977739-6980501 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp2 | NM_001166368.1 | 2 | 137 | NP_001159840.1 |
| NM_008020.3 | 2 | 152 | NP_032046.1 | |
| AK144284.1 | 2 | 153 | BAE25814.1 | |
| AK165394.1 | 2 | 117 | BAE38158.1 | |
| BC031824.1 | 2 | 106 | AAH31824.1 | |
| BC053692.1 | 2 | 142 | AAH53692.1 | |
| DV655383.1 | 2 | 137 |

Genomic Map