Assay Details
Target Gene Details
Entrez Gene ID: | 240549 |
Gene Name: | predicted gene 4952 |
Gene Aliases: |
EG240549 |
Location: |
Chr.19:12599984-12627616 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm4952 | NM_001013762.2 | NP_001013784.2 | ||
| NM_001167907.1 | NP_001161379.1 | |||
| BC089619.1 | AAH89619.1 |
More Information
Set Membership: |
|

Genomic Map