Assay Details
Target Gene Details
Entrez Gene ID: | 73707 |
Gene Name: | guanylate cyclase 2g |
Gene Aliases: |
2410077I05Rik, GC-G |
Location: |
Chr.19:55198098-55242429 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy2g | NM_001081076.2 | NP_001074545.1 | ||
| AY395631.1 | AAR28089.1 |
More Information
Set Membership: |
|

Genomic Map