Assay Details
Target Gene Details
Entrez Gene ID: | 56072 |
Gene Name: | lectin, galactose binding, soluble 12 |
Gene Aliases: |
AW987437, Grip1 |
Location: |
Chr.19:7596656-7610466 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgals12 | NM_019516.3 | 3 | 713 | NP_062389.1 |
| AF223223.1 | 4 | 407 | AAF34682.1 | |
| AF244978.1 | 4 | 415 | AAK77326.1 | |
| AF244979.1 | 3 | 460 | AAK77327.1 | |
| AK033535.1 | 3 | 640 | BAC28345.1 | |
| AK085660.1 | 3 | 716 | BAC39498.1 | |
| AK136526.1 | 3 | 714 | BAE23026.1 | |
| BC030890.1 | 3 | 397 | AAH30890.1 |
More Information
Set Membership: |
|

Genomic Map