Assay Details
Target Gene Details
Entrez Gene ID: | 56072 |
Gene Name: | lectin, galactose binding, soluble 12 |
Gene Aliases: |
AW987437, Grip1 |
Location: |
Chr.19:7596656-7610466 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgals12 | NM_019516.3 | NP_062389.1 | ||
| AF223223.1 | AAF34682.1 | |||
| AF244978.1 | AAK77326.1 | |||
| AF244979.1 | AAK77327.1 | |||
| AK033535.1 | BAC28345.1 | |||
| AK085660.1 | BAC39498.1 | |||
| AK136526.1 | BAE23026.1 | |||
| BC030890.1 | AAH30890.1 |
More Information
Set Membership: |
|

Genomic Map