Assay Details
Target Gene Details
Entrez Gene ID: | 13041 |
Gene Name: | cathepsin W |
Gene Aliases: |
lymphopain |
Location: |
Chr.19:5465240-5470387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsw | NM_009985.4 | NP_034115.2 | ||
| AF014941.1 | AAB82455.1 | |||
| AI450305.1 | ||||
| AK088375.1 | BAC40314.1 | |||
| AK163626.1 | ||||
| BE447240.1 |
More Information
Set Membership: |
|

Genomic Map