Assay Details
Target Gene Details
Entrez Gene ID: | 70387 |
Gene Name: | tetratricopeptide repeat domain 9C |
Gene Aliases: |
2210019E14Rik, 6330408J23Rik |
Location: |
Chr.19:8809066-8819780 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc9c | NM_027412.3 | 2 | 316 | NP_081688.2 |
| AK008753.1 | 1 | 647 | BAB25875.1 | |
| AK018145.1 | 2 | 357 | ||
| AK169288.1 | 2 | 317 | BAE41045.1 | |
| AK169296.1 | 2 | 317 | BAE41052.1 | |
| BC043061.1 | 1 | 628 | AAH43061.1 | |
| BC058967.1 | 1 | 628 | AAH58967.1 | |
| BC063773.1 | 1 | 282 | AAH63773.1 |
More Information
Set Membership: |
|

Genomic Map