Assay Details
Target Gene Details
Entrez Gene ID: | 225845 |
Gene Name: | phospholipase A2, group XVI |
Gene Aliases: |
C78643, HRSL3, Hrasls3, Hrev107, MLP-3 |
Location: |
Chr.19:7557434-7588545 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g16 | NM_139269.2 | NP_644675.2 | ||
| AK039703.1 | BAE20550.1 | |||
| AK042007.1 | BAE20615.1 | |||
| AK052657.1 | BAC35085.1 | |||
| AK163505.1 | BAE37375.1 | |||
| AX775891.1 | CAE11645.1 | |||
| BC024581.1 | AAH24581.1 |

Genomic Map