Assay Details
Target Gene Details
Entrez Gene ID: | 17476 |
Gene Name: | macrophage expressed gene 1 |
Gene Aliases: |
MPS1, Mpg-1 |
Location: |
Chr.19:12460709-12465285 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpeg1 | NM_010821.1 | NP_034951.1 | ||
| AK030216.1 |

Genomic Map