Assay Details
Target Gene Details
Entrez Gene ID: | 68642 |
Gene Name: | transmembrane protein 216 |
Gene Aliases: |
1110017C22Rik, 2810441K11Rik, 4921533J23Rik, A930021F15Rik, AI482550 |
Location: |
Chr.19:10539326-10556303 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem216 | NM_026798.3 | 1 | 63 | NP_081074.1 |
| AK003740.1 | 1 | 65 | BAB22973.1 | |
| BY704000.1 | 1 | 65 |

Genomic Map