Assay Details
Target Gene Details
Entrez Gene ID: | 67710 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide G |
Gene Aliases: |
2410046K11Rik, A230108L04Rik, C76415, RBP7, Rpo2-7l |
Location: |
Chr.19:8793129-8798557 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2g | NM_026329.2 | NP_080605.1 | ||
| AK007892.1 | ||||
| AK010687.1 | BAB27119.1 | |||
| AK011627.1 | BAB27743.1 | |||
| AK013134.1 | BAB28670.1 | |||
| AK017473.1 | BAB30760.1 | |||
| BC005580.1 | AAH05580.3 | |||
| BC055278.1 | AAH55278.1 |
More Information
Set Membership: |
|

Genomic Map