Assay Details
Target Gene Details
Entrez Gene ID: | 70999 |
Gene Name: | N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae) |
Gene Aliases: |
4931433E08Rik, AU023197, Nat11 |
Location: |
Chr.19:7225668-7241222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa40 | NM_027643.1 | NP_081919.1 | ||
| AK016504.1 | BAB30275.1 | |||
| AK033510.1 | ||||
| AK033896.1 | BAC28506.1 | |||
| AK035418.1 | BAE20496.1 | |||
| AK162625.1 | ||||
| AK170548.1 | BAE41873.1 | |||
| AK202080.1 | ||||
| AK209446.1 | ||||
| AK218786.1 | 1 | 164 | ||
| BC020020.1 | AAH20020.1 | |||
| BC058212.1 | 1 | 349 | AAH58212.1 |

Genomic Map