Assay Details
Target Gene Details
Entrez Gene ID: | 102902673 |
Gene Name: | predicted gene 21992 |
Gene Aliases: |
Rbm14-rbm4 |
Location: |
Chr.19:4784293-4811634 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21992 | NM_001290127.1 | NP_001277056.1 | ||
| NM_001290128.1 | NP_001277057.1 | |||
| AK087488.1 | 2 | 586 | BAC39895.1 | |
| EU287940.1 | ||||
| EU287941.1 |
Target Gene Details
Entrez Gene ID: | 19653 |
Gene Name: | RNA binding motif protein 4 |
Gene Aliases: |
4921506I22Rik, Lark1, Mlark, Rbm4a, lark |
Location: |
Chr.19:4784293-4794009 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm4 | NM_001290122.1 | NP_001277051.1 | ||
| NM_001290123.1 | NP_001277052.1 | |||
| NM_001290124.1 | NP_001277053.1 | |||
| NM_001290125.1 | 1 | 45 | NP_001277054.1 | |
| NM_009032.3 | 1 | 45 | NP_033058.2 | |
| AK032454.1 | BAE20479.1 | |||
| AK049659.1 | BAC33862.1 | |||
| AK158962.1 | BAE34743.1 | |||
| AK182159.1 | ||||
| AK182573.1 | ||||
| BC090633.1 | AAH90633.1 | |||
| BC144949.1 | ||||
| BC144950.1 | ||||
| BY261809.1 | 1 | 46 | ||
| CK331622.1 | ||||
| CN670081.1 | ||||
| DV660645.1 | ||||
| U89506.1 | AAC53171.1 |
More Information
Set Membership: |
|

Genomic Map