Assay Details
Target Gene Details
Entrez Gene ID: | 17995 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 1 |
Gene Aliases: |
CI-51kD |
Location: |
Chr.19:4007499-4012755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufv1 | NM_133666.3 | NP_598427.1 | ||
| AK005021.1 | ||||
| AK075692.1 | BAC35893.1 | |||
| AK151455.1 | ||||
| AK187072.1 | ||||
| AK189391.1 | ||||
| AK206448.1 | ||||
| BC014818.1 | AAH14818.1 | |||
| BC041682.1 | AAH41682.1 | |||
| BE863504.1 |
More Information
Set Membership: |
|

Genomic Map