Assay Details
Target Gene Details
Entrez Gene ID: | 18552 |
Gene Name: | proprotein convertase subtilisin/kexin type 5 |
Gene Aliases: |
PC5, PC5/6A, PC5A, PC6, SPC6, b2b1549Clo, b2b585Clo |
Location: |
Chr.19:17432832-17837632 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcsk5 | NM_001163144.1 | NP_001156616.1 | ||
| NM_001190483.1 | NP_001177412.1 | |||
| AK143998.1 | ||||
| BC013068.1 | AAH13068.1 | |||
| D12619.1 | BAA02143.1 | |||
| D17583.1 | BAA04507.1 | |||
| L14932.1 | AAA74636.1 |
More Information
Set Membership: |
|

Genomic Map