Assay Details
Target Gene Details
Entrez Gene ID: | 18719 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase, type 1 beta |
Gene Aliases: |
PI4P5K-I[b], PIP5K1-beta, PIP5KIbeta, Pipk5b, STM7 |
Location: |
Chr.19:24294794-24555827 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip5k1b | NM_008846.2 | 6 | 547 | NP_032872.1 |
| AF048695.1 | 4 | 396 | AAC05374.1 | |
| AK133809.1 | 6 | 542 | ||
| AK146100.1 | 6 | 547 | BAE26901.1 | |
| AK149354.1 | 6 | 547 | BAE28830.1 | |
| AK164552.1 | 6 | 530 | BAE37836.1 | |
| BC034864.1 | 6 | 561 | AAH34864.1 | |
| D86176.1 | 5 | 480 | BAA13030.1 |
More Information
Set Membership: |
|

Genomic Map