Assay Details
Target Gene Details
Entrez Gene ID: | 17995 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 1 |
Gene Aliases: |
CI-51kD |
Location: |
Chr.19:4007499-4012755 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufv1 | NM_133666.3 | 7 | 1137 | NP_598427.1 |
| AK005021.1 | 7 | 1085 | ||
| AK075692.1 | 7 | 1082 | BAC35893.1 | |
| AK151455.1 | 7 | 1048 | ||
| AK178320.1 | 3 | 325 | ||
| AK185238.1 | 3 | 327 | ||
| AK187850.1 | 3 | 325 | ||
| AK188080.1 | 2 | 220 | ||
| AK188929.1 | 2 | 207 | ||
| AK189786.1 | 2 | 326 | ||
| AK205680.1 | 2 | 198 | ||
| AK209435.1 | 3 | 325 | ||
| AK210493.1 | ||||
| AK218408.1 | 3 | 324 | ||
| AW113957.1 | ||||
| BC014818.1 | 7 | 1107 | AAH14818.1 | |
| BC041682.1 | 7 | 1040 | AAH41682.1 |
More Information
Set Membership: |
|

Genomic Map