Assay Details
Target Gene Details
Entrez Gene ID: | 56078 |
Gene Name: | carbonic anhydrase 5b, mitochondrial |
Gene Aliases: |
7330410H16Rik, CAVB, Ca5b, CarVb, D730005F19Rik |
Location: |
Chr.X:163976822-164028010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car5b | NM_181315.4 | NP_851832.2 | ||
| AF192978.1 | AAF08291.1 | |||
| AK085670.1 | BAC39501.1 | |||
| BC034413.1 | AAH34413.1 |

Genomic Map