Assay Details
Target Gene Details
Entrez Gene ID: | 109113 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains 2 |
Gene Aliases: |
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf |
Location: |
Chr.19:30030433-30093729 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uhrf2 | NM_144873.2 | 11 | 1950 | NP_659122.2 |
| AB116653.1 | 11 | 1685 | BAC81739.1 | |
| AF274047.1 | 11 | 1860 | AAM33798.1 | |
| AK170648.1 | 2 | 106 | BAE41935.1 | |
| BC060241.1 | 11 | 1950 | AAH60241.1 |
More Information
Set Membership: |
|

Genomic Map