Assay Details
Target Gene Details
Entrez Gene ID: | 668257 |
Gene Name: | diacylglycerol O-acyltransferase 2-like 6 |
Gene Aliases: |
9930021F22Rik |
Location: |
Chr.X:100516094-100546108 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgat2l6 | NM_001114084.1 | NP_001107556.1 | ||
| AK079438.1 |
More Information
Set Membership: |
|

Genomic Map