Assay Details
Target Gene Details
Entrez Gene ID: | 54632 |
Gene Name: | FtsJ homolog 1 (E. coli) |
Gene Aliases: |
AI931847, Ftsj, Ftsjl, Sfc12 |
Location: |
Chr.X:8238668-8252406 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ftsj1 | NM_001290430.1 | NP_001277359.1 | ||
| NM_133991.2 | NP_598752.2 | |||
| AK004357.1 | ||||
| AK036313.1 | BAC29380.1 | |||
| AK150251.1 | 3 | 274 | ||
| BC011144.1 | AAH11144.1 | |||
| BC034880.1 | AAH34880.1 |
More Information
Set Membership: |
|

Genomic Map