Assay Details
Target Gene Details
Entrez Gene ID: | 72997 |
Gene Name: | Kdm5c adjacent non-coding transcript |
Gene Aliases: |
2810454L23Rik, 2900056M20Rik, Spasm |
Location: |
Chr.X:152294824-152327493 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kantr | NR_040269.2 | |||
| NR_040270.2 | ||||
| AK013706.1 | 1 | 319 | ||
| AW213569.1 | ||||
| BB852892.1 | ||||
| BY201328.1 |

Genomic Map