Assay Details
Target Gene Details
Entrez Gene ID: | 14381 |
Gene Name: | glucose-6-phosphate dehydrogenase X-linked |
Gene Aliases: |
G28A, G6pd, Gpdx |
Location: |
Chr.X:74409486-74429161 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6pdx | NM_008062.2 | NP_032088.1 | ||
| AK088135.1 | BAC40166.1 | |||
| AK148031.1 | ||||
| AK165207.1 | BAE38077.1 | |||
| BC075663.1 | AAH75663.1 | |||
| Z11911.1 | CAA77967.1 |
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkg | NM_001136067.2 | NP_001129539.1 | ||
| NM_001161421.1 | NP_001154893.1 | |||
| NM_001161423.1 | NP_001154895.1 | |||
| NM_001161424.1 | NP_001154896.1 | |||
| AF513109.1 | AAP47160.1 | |||
| AY112937.1 | AAM54725.1 | |||
| BC021431.1 | AAH21431.1 | |||
| CJ099503.1 | ||||
| CN717929.1 | ||||
| CV556906.1 |
More Information
Set Membership: |
|

Genomic Map