Assay Details
Target Gene Details
Entrez Gene ID: | 59048 |
Gene Name: | C1GALT1-specific chaperone 1 |
Gene Aliases: |
1500002I11Rik, AB030184, C1GalT2, C81205, Cosmc |
Location: |
Chr.X:38630783-38635143 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1galt1c1 | NM_021550.3 | 1 | 96 | NP_067525.1 |
| AB030184.1 | BAA92748.1 | |||
| AK005118.1 | ||||
| AK008040.1 | BAB25426.1 | |||
| AK161729.1 | BAE36551.1 | |||
| AY159320.1 | AAN78130.1 | |||
| BC029909.1 | 1 | 62 | AAH29909.1 |
More Information
Set Membership: |
|

Genomic Map