Assay Details
Target Gene Details
Entrez Gene ID: | 22589 |
Gene Name: | alpha thalassemia/mental retardation syndrome X-linked homolog (human) |
Gene Aliases: |
4833408C14Rik, AI447451, ATR2, DXHXS6677E, HP1-BP38, Hp1bp2, Hp1bp38, MRXS3, RAD54L, Rad54, XH2, Xnp, ZNF-HX |
Location: |
Chr.X:105797615-105929408 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atrx | NM_009530.2 | NP_033556.2 | ||
| AF026032.1 | AAC08741.1 | |||
| AK082779.1 | 6 | 1273 | ||
| AK134024.1 | BAE21982.1 | |||
| AK161784.1 | BAE36571.1 |
More Information
Set Membership: |
|

Genomic Map