Assay Details
Target Gene Details
Entrez Gene ID: | 74448 |
Gene Name: | ADP-ribosylation factor-like 13A |
Gene Aliases: |
4933422M21Rik, Arl13 |
Location: |
Chr.X:134187501-134208030 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl13a | NM_028947.1 | NP_083223.1 | ||
| AK016872.1 | BAB30477.1 | |||
| BC100522.1 | AAI00523.1 |

Genomic Map