Assay Details
Target Gene Details
Entrez Gene ID: | 382253 |
Gene Name: | cyclin-dependent kinase-like 5 |
Gene Aliases: |
BC038161, Stk9 |
Location: |
Chr.X:160784308-160994681 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkl5 | NM_001024624.2 | NP_001019795.1 | ||
| AK052380.1 | BAC34965.1 | |||
| AK139220.1 | BAE23922.1 | |||
| BY291331.1 |
Target Gene Details
Entrez Gene ID: | 414089 |
Gene Name: | gap junction protein, alpha 6 |
Gene Aliases: |
A530015O12Rik, Cnx33, Cx33, Gja-6 |
Location: |
Chr.X:160902116-160907052 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja6 | NM_001001496.2 | 1 | 51 | NP_001001496.1 |
| AK040693.1 | BAC30669.1 | |||
| AK146838.1 | 1 | 51 | BAE27472.1 | |
| AK168953.1 | 1 | 51 | BAE40759.1 | |
| BC117760.2 |
More Information
Set Membership: |
|

Genomic Map