Assay Details
Target Gene Details
Entrez Gene ID: | 245532 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 2 |
Gene Aliases: |
9430062J17Rik, ARAT, Dgat2l4, WS |
Location: |
Chr.X:100399336-100494221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Awat2 | NM_001290395.1 | NP_001277324.1 | ||
| NM_177746.4 | NP_808414.2 | |||
| AK034920.1 | BAC28882.1 | |||
| AY611031.1 | AAT68765.1 | |||
| AY611032.1 | AAT68766.1 | |||
| BC138085.1 | ||||
| BC138086.1 |
More Information
Set Membership: |
|

Genomic Map