Assay Details
Target Gene Details
Entrez Gene ID: | 18700 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class A |
Gene Aliases: |
AI194334, Pig-a |
Location: |
Chr.X:164419680-164433916 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Piga | NM_011081.2 | NP_035211.2 | ||
| AK032283.1 | BAC27792.1 | |||
| AK039802.1 | ||||
| AK161026.1 | BAE36154.1 | |||
| AK171453.1 | BAE42462.1 | |||
| BY270710.1 | ||||
| D26047.1 | BAA05047.1 | |||
| S78188.1 | AAP32009.1 |
More Information
Set Membership: |
|

Genomic Map