Assay Details
Target Gene Details
Entrez Gene ID: | 93761 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 |
Gene Aliases: |
5730494M04Rik, Snf2l |
Location: |
Chr.X:47809370-47892931 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarca1 | NM_001290708.1 | 16 | 2288 | NP_001277637.1 |
| NM_053123.4 | 16 | 2288 | NP_444353.3 | |
| AF325920.1 | 16 | 2295 | AAK52453.1 | |
| AK030741.1 | 16 | 2255 | BAC27109.1 | |
| AK032978.1 | ||||
| BC016071.1 | 16 | 2265 | ||
| BC057115.1 | 16 | 2288 | AAH57115.1 |
More Information
Set Membership: |
|

Genomic Map