Assay Details
Target Gene Details
Entrez Gene ID: | 20523 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, brain), member 14 |
Gene Aliases: |
BMCP1, UCP5, UCP5L, UCP5S |
Location: |
Chr.X:48623418-48662298 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a14 | NM_001166450.2 | 8 | 976 | NP_001159922.1 |
| NM_001290703.1 | 10 | 1165 | NP_001277632.1 | |
| NM_001290704.1 | 10 | 1156 | NP_001277633.1 | |
| NM_001290705.1 | 7 | 731 | NP_001277634.1 | |
| NM_011398.3 | 8 | 1063 | NP_035528.1 | |
| AF076981.1 | 8 | 1064 | AAD03674.1 | |
| AF155812.1 | 7 | 651 | AAG29585.1 | |
| AF155813.1 | 7 | 642 | AAG29586.1 | |
| AF535150.1 | 10 | 1157 | AAN63885.1 | |
| AF535151.1 | 10 | 1166 | AAN63886.1 | |
| AK148277.1 | 8 | 2511 | ||
| BC048692.1 | 7 | 786 | AAH48692.1 |
More Information
Set Membership: |
|

Genomic Map