Assay Details
Target Gene Details
Entrez Gene ID: | 20937 |
Gene Name: | suppressor of variegation 3-9 homolog 1 (Drosophila) |
Gene Aliases: |
AI852103, AL022883, DXHXS7466e, H3-K9-HMTase 1, KMT1A, mIS6 |
Location: |
Chr.X:8061171-8074760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suv39h1 | NM_001290716.1 | NP_001277645.1 | ||
| NM_011514.2 | NP_035644.1 | |||
| AF019969.1 | AAB92225.1 | |||
| AF193861.1 | AAF60969.1 | |||
| AF193862.1 | AAF60970.1 | |||
| AK088405.1 | BAC40334.1 | |||
| AK132238.1 | 4 | 1007 | ||
| AK169389.1 | BAE41136.1 | |||
| AK169592.1 | ||||
| BC023860.1 | AAH23860.1 |
More Information
Set Membership: |
|

Genomic Map