Assay Details
Target Gene Details
Entrez Gene ID: | 22284 |
Gene Name: | ubiquitin specific peptidase 9, X chromosome |
Gene Aliases: |
5730589N07Rik, AA407302, AA407699, AL022658, AL022749, Dffrx, FAF-X, Fafl |
Location: |
Chr.X:13070997-13173328 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp9x | NM_009481.2 | NP_033507.2 | ||
| AK028443.1 | ||||
| AK049283.1 | BAC33657.2 | |||
| DQ086491.1 | AAY99447.1 | |||
| U67874.1 | AAB07731.1 |
More Information
Set Membership: |
|

Genomic Map