Assay Details
Target Gene Details
Entrez Gene ID: | 26900 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked |
Gene Aliases: |
8030469F12Rik, D1Pas1-rs1, Dby |
Location: |
Chr.Y:1260715-1286628 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx3y | NM_012008.2 | NP_036138.1 | ||
| AJ007376.1 | CAA07483.1 | |||
| AK154382.1 | BAE32549.1 | |||
| BC021453.1 | AAH21453.1 | |||
| CD552910.1 |
More Information
Set Membership: |
|

Genomic Map