Assay Details
Target Gene Details
Entrez Gene ID: | 407786 |
Gene Name: | TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
BC066223, Taf9l |
Location: |
Chr.X:106206874-106221158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf9b | NM_001001176.2 | NP_001001176.2 | ||
| NM_001167988.1 | NP_001161460.1 | |||
| AK131710.1 | BAE20773.1 | |||
| AK135097.1 | BAE22420.1 | |||
| BC051635.1 | AAH51635.1 | |||
| BC066223.1 | AAH66223.1 |

Genomic Map