Assay Details
Target Gene Details
Entrez Gene ID: | 66226 |
Gene Name: | trafficking protein particle complex 2 |
Gene Aliases: |
1110066L09Rik, 1810064C02Rik, AW496358, MIP-2A, Sedl, TRS20 |
Location: |
Chr.X:166440825-166452543 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc2 | NM_025432.3 | NP_079708.2 | ||
| AK002658.1 | BAB22265.1 | |||
| AK007889.1 | BAB25332.1 | |||
| AK008564.1 | BAB25747.1 | |||
| AK034967.1 | 3 | 1090 |
More Information
Set Membership: |
|

Genomic Map