Assay Details
Target Gene Details
Entrez Gene ID: | 56805 |
Gene Name: | zinc finger and BTB domain containing 33 |
Gene Aliases: |
AW260253, E130014G12Rik, Kaiso, Znf-kaiso |
Location: |
Chr.X:38189793-38197046 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb33 | NM_001079513.1 | NP_001072981.1 | ||
| NM_020256.2 | NP_064652.2 | |||
| AF097416.1 | AAD20989.1 | |||
| AK087417.1 | BAC39866.1 | |||
| AK089423.1 | BAC40875.1 | |||
| BY200826.1 |
More Information
Set Membership: |
|

Genomic Map